2-Chlor-4-ethoxyphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chloro-4-ethoxybenzeneboronic acid
OR13119-5G
APOSOR13119-5GEA
105
EUR
APOSOR13119-5G
2-Chlor-4-ethoxyphenylboronsäure 98%
2-Chlor-4-ethoxyphenylboronsäure
Formel:
C₈H₁₀BClO₃ Molecular Weight: 200,43 g/mol Schmelzpunkt: 104…108 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02684318 CAS-Nummer: 313545-44-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...