5-Chlor-2-ethoxyphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Chloro-2-ethoxybenzeneboronic acid
OR13120-5G
OR13120-25G
APOSOR13120-5GEA
36.4
EUR
APOSOR13120-5G
APOSOR13120-25G
5-Chlor-2-ethoxyphenylboronsäure 98%
5-Chlor-2-ethoxyphenylboronsäure
Formel:
C₈H₁₀BClO₃ Schmelzpunkt: 120…124 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03427055 CAS-Nummer: 352534-86-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...