3-(tert-Butylcarbamoyl)phenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(tert-Butylaminocarbonyl)phenylboronic acid
OR13122-1G
OR13122-5G
APOSOR13122-1GEA
116
EUR
APOSOR13122-1G
APOSOR13122-5G
3-(tert-Butylcarbamoyl)phenylboronsäure
3-(tert-Butylcarbamoyl)phenylboronsäure
Formel:
C₁₁H₁₆BNO₃ Schmelzpunkt: 120…126 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04115693 CAS-Nummer: 183158-30-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...