3-(2,3-Dihydro-1,4-benzodioxin-6-yl)acrylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR13151-1G
OR13151-5G
APOSOR13151-1GEA
336
EUR
APOSOR13151-1G
APOSOR13151-5G
3-(2,3-Dihydro-1,4-benzodioxin-6-yl)acrylic acid
3-(2,3-Dihydro-1,4-benzodioxin-6-yl)acrylic acid
Formel:
C₁₁H₁₀O₄ Schmelzpunkt: 196…198 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00168684 CAS-Nummer: 14939-91-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...