Thiazole-4-carboxaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Thiazole-4-carbaldehyde
OR13153-1G
OR13153-500MG
OR13153-5G
APOSOR13153-1GEA
43.5
EUR
APOSOR13153-1G
APOSOR13153-500MG
APOSOR13153-5G
Thiazole-4-carboxaldehyde
Thiazole-4-carboxaldehyde
Formel:
C₄H₃NOS Molecular Weight: 113,13 g/mol Schmelzpunkt: 61…65 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00626896 CAS-Nummer: 3364-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...