3-(3-Hydroxypropyl)benzeneboronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-(3-Hydroxypropyl)phenyl)boronic acid

OR13154-100MG
APOSOR13154-100MGEA 62.5 EUR
APOSOR13154-100MG
3-(3-Hydroxypropyl)benzeneboronic acid
3-(3-Hydroxypropyl)benzeneboronic acid
Formel: C₉H₁₃BO₃
Molecular Weight: 180,01 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04115650
CAS-Nummer: 736989-98-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR