3-(2-Methoxycarbonylethyl)benzeneboronic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-(3-Methoxy-3-oxopropyl)phenyl)boronic acid
OR13161-1G
OR13161-5G
APOSOR13161-1GEA
162
EUR
APOSOR13161-1G
APOSOR13161-5G
3-(2-Methoxycarbonylethyl)benzeneboronic acid 98%
3-(2-Methoxycarbonylethyl)benzeneboronic acid
Formel:
C₁₀H₁₃BO₄ Molecular Weight: 208.02 g/mol Schmelzpunkt: 96…100 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04115648 CAS-Nummer: 833472-82-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...