3-(2-Carboxyethyl)phenylboronsäure 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(2-Carboxyethyl)benzeneboronic acid, 3-(3-Boronophenyl)propanoic acid
OR13162-1G
OR13162-5G
APOSOR13162-1GEA
224
EUR
APOSOR13162-1G
APOSOR13162-5G
3-(2-Carboxyethyl)phenylboronsäure 96%
3-(2-Carboxyethyl)phenylboronsäure
Formel:
C₉H₁₁BO₄ Molecular Weight: 193,99 g/mol Schmelzpunkt: 166…169 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115649 CAS-Nummer: 693803-17-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...