3-(2-Carboxyvinyl)phenylboronsäure 97+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(3-Boronophenyl)acrylic acid, 3-(2-Carboxyvinyl)benzeneboronic acid
OR13173-1G
OR13173-5G
APOSOR13173-1GEA
79.9
EUR
APOSOR13173-1G
APOSOR13173-5G
3-(2-Carboxyvinyl)phenylboronsäure 97+%
3-(2-Carboxyvinyl)phenylboronsäure
Formel:
C₉H₉BO₄ Molecular Weight: 191,98 g/mol Schmelzpunkt: 218 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01075738 CAS-Nummer: 216144-91-1 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...