4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzoic acid

Lieferant: Apollo Scientific
Achtung

OR13183-1G OR13183-5G
APOSOR13183-1GEA 120 EUR
APOSOR13183-1G APOSOR13183-5G
4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzoic acid
4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzoic acid
Formel: C₁₂H₁₂N₂O₂
Molecular Weight: 216.24 g/mol
Schmelzpunkt: 160…163 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03011525
CAS-Nummer: 81282-82-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR