ω-Bromacetophenon 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: α-Bromacetophenon, Phenacylbromid, 2-Bromacetophenon

OR13196-100G OR13196-25G
APOSOR13196-100GEA 87 EUR
APOSOR13196-100G APOSOR13196-25G
ω-Bromacetophenon 97%
ω-Bromacetophenon
Formel: (C₆H₅)C(O)CH₂Br
Molecular Weight: 199,05 g/mol
Schmelzpunkt: 48…51 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00000195
CAS-Nummer: 70-11-1
EINECS: 200-724-9
UN: 2645
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR