2-Brom-5-chloranisol

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-4-chloro-2-methoxybenzene

OR13198-25G OR13198-5G OR13198-100G
APOSOR13198-25GEA 29 EUR
APOSOR13198-25G APOSOR13198-5G APOSOR13198-100G
2-Brom-5-chloranisol
2-Brom-5-chloranisol
Formel: C₇H₆BrClO
Molecular Weight: 221,48 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03790889
CAS-Nummer: 174913-09-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR