4-Amino-3-nitropyridin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Nitropyridin-4-amine, 3-Nitro-4-pyridylamine, 3-Nitropyridin-4-ylamine
OR13224-25G
OR13224-5G
APOSOR13224-25GEA
42.2
EUR
APOSOR13224-25G
APOSOR13224-5G
4-Amino-3-nitropyridin
4-Amino-3-nitropyridin
Formel:
C₅H₅N₃O₂ Molecular Weight: 139,11 g/mol Schmelzpunkt: 200…207 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00160304 CAS-Nummer: 1681-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...