3-Ethoxy-4-hydroxy-5-nitrobenzaldehyd

Lieferant: Apollo Scientific
Achtung

OR13238-1G OR13238-250MG
APOSOR13238-1GEA 29 EUR
APOSOR13238-1G APOSOR13238-250MG
3-Ethoxy-4-hydroxy-5-nitrobenzaldehyd
3-Ethoxy-4-hydroxy-5-nitrobenzaldehyde
Formel: C₉H₉NO₅
Molecular Weight: 211,1715 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00058678
CAS-Nummer: 178686-24-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR