6-Brompiperonal

Lieferant: Apollo Scientific

Synonyme: 6-Brom-1,3-benzodioxol-5-carbaldehyd

OR13261-100G OR13261-25G
APOSOR13261-100GEA 324 EUR
APOSOR13261-100G APOSOR13261-25G
6-Brompiperonal
6-Brompiperonal
Formel: C₈H₅BrO₃
Molecular Weight: 229.03 g/mol
Schmelzpunkt: 125…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00022952
CAS-Nummer: 15930-53-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR