3-Amino-3-(4-bromphenyl)propionsäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR13344-1G
OR13344-5G
APOSOR13344-1GEA
21.9
EUR
APOSOR13344-1G
APOSOR13344-5G
3-Amino-3-(4-bromphenyl)propionsäure 95%
3-Amino-3-(4-bromphenyl)propionsäure
Formel:
C₉H₁₀BrNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00187112 CAS-Nummer: 39773-47-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...