Bzl-Pro-OH

Lieferant: Apollo Scientific
Achtung

Synonyme: (2S)-1-Benzylpyrrolidine-2-carboxylic acid, (S)-1-Benzylpyrrolidine-2-carboxylic acid

OR13357-1G OR13357-5G
APOSOR13357-1GEA 58 EUR
APOSOR13357-1G APOSOR13357-5G
Bzl-Pro-OH
Bzl-Pro-OH
Formel: C₁₂H₁₅NO₂
Molecular Weight: 205,3 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00235894
CAS-Nummer: 31795-93-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR