8-Amino-6-bromchinolin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromoquinolin-8-amine

OR13403-250MG
APOSOR13403-250MGEA 71.2 EUR
APOSOR13403-250MG
8-Amino-6-bromchinolin 97%
8-Amino-6-bromoquinoline
Formel: C₉H₇BrN₂
Molecular Weight: 223,0700 g/mol
Schmelzpunkt: 197 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00194777
CAS-Nummer: 57339-57-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR