2-Chloro-N-phenethylacetamide 98%

Lieferant: Apollo Scientific
Achtung

OR13488-1G OR13488-5G
APOSOR13488-1GEA 63.8 EUR
APOSOR13488-1G APOSOR13488-5G
2-Chloro-N-phenethylacetamide 98%
2-Chloro-N-phenethylacetamide
Formel: C₁₀H₁₂ClNO
Molecular Weight: 197.67 g/mol
Schmelzpunkt: 67…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00245084
CAS-Nummer: 13156-95-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR