2-Chinolincarbonitril 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Quinoline-2-carbonitrile
OR13512-1G
OR13512-5G
OR13512-25G
APOSOR13512-1GEA
21.9
EUR
APOSOR13512-1G
APOSOR13512-5G
APOSOR13512-25G
2-Chinolincarbonitril 97%
2-Chinolincarbonitril
Formel:
C₁₀H₆N₂ Molecular Weight: 154,17 g/mol Siedepunkt: 160 °C/23mmHg Schmelzpunkt: 93…95 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134341 CAS-Nummer: 1436-43-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...