2-(Methoxymethyl)pyridine

Lieferant: Apollo Scientific
Gefahr

OR13609-1G OR13609-5G
APOSOR13609-1GEA 119 EUR
APOSOR13609-1G APOSOR13609-5G
2-(Methoxymethyl)pyridine
2-(Methoxymethyl)pyridine
Formel: C₇H₉NO
Molecular Weight: 123.1500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00030825
CAS-Nummer: 23579-92-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR