4-Chlor-3-nitrozimtsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(4-Chloro-3-nitrophenyl)prop-2-enoic acid, 3-(4-Chloro-3-nitrophenyl)acrylic acid
OR1365-25G
OR1365-5G
APOSOR1365-25GEA
109
EUR
APOSOR1365-25G
APOSOR1365-5G
4-Chlor-3-nitrozimtsäure 98%
4-Chlor-3-nitrozimtsäure
Formel:
C₉H₆ClNO₄ Molecular Weight: 227,601 g/mol Schmelzpunkt: 188…190 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063311 CAS-Nummer: 20797-48-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...