2-Bromcarbazol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-9H-carbazole

OR13661-1G OR13661-5G OR13661-25G
APOSOR13661-1GEA 21.9 EUR
APOSOR13661-1G APOSOR13661-5G APOSOR13661-25G
2-Bromcarbazol
2-Bromocarbazole
Formel: C₁₂H₈BrN
Molecular Weight: 246,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09750430
CAS-Nummer: 3652-90-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR