3-Brom-1H-1,2,4-triazol

Lieferant: Apollo Scientific
Achtung

OR13675-5G OR13675-25G
APOSOR13675-5GEA 21.9 EUR
APOSOR13675-5G APOSOR13675-25G
3-Brom-1H-1,2,4-triazol
3-Brom-1H-1,2,4-triazol
Formel: C₂H₂BrN₃
Molecular Weight: 147.96 g/mol
Schmelzpunkt: 187…189 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01421960
CAS-Nummer: 7343-33-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR