2,6-Dibromo-5-ethoxypyridin-3-amine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Amino-2,6-dibromo-5-ethoxypyridine
OR13676-1G
OR13676-250MG
OR13676-5G
APOSOR13676-1GEA
72.5
EUR
APOSOR13676-1G
APOSOR13676-250MG
APOSOR13676-5G
2,6-Dibromo-5-ethoxypyridin-3-amine
2,6-Dibromo-5-ethoxypyridin-3-amine
Formel:
C₇H₈Br₂N₂O Lagertemperatur: Raumtemperatur |
CAS-Nummer:
1000018-10-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...