Methyl-3-iod-4-methylbenzoat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 3-iodo-4-methylbenzoate
OR13678-100G
OR13678-25G
OR13678-10G
APOSOR13678-100GEA
92.8
EUR
APOSOR13678-100G
APOSOR13678-25G
APOSOR13678-10G
Methyl-3-iod-4-methylbenzoat
Methyl-3-iod-4-methylbenzoat
Formel:
C₉H₉IO₂ Molecular Weight: 276,07 g/mol Schmelzpunkt: 26…29 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00230583 CAS-Nummer: 90347-66-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...