Idebenon 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(10-Hydroxydecyl)-5,6-dimethoxy-3-methyl-1,4-benzoquinone, 2-(10-Hydroxydecyl)-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione
OR13682-1G
OR13682-5G
OR13682-25G
APOSOR13682-1GEA
51
EUR
APOSOR13682-1G
APOSOR13682-5G
APOSOR13682-25G
Idebenon 98%
Idebenone
Formel:
C₁₉H₃₀O₅ Molecular Weight: 338,44 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00274552 CAS-Nummer: 58186-27-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...