2-(3-Ethoxy-4-ethoxycarbonylphenyl)acetic acid

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-Carboxymethyl-2-ethoxybenzoic Acid Ethyl Ester, Ethyl 4-Carboxymethyl-2-ethoxybenzoate, 2-(3-Ethoxy-4-(ethoxycarbonyl)phenyl)acetic acid

OR13687-25G
APOSOR13687-25GEA 68.3 EUR
APOSOR13687-25G
2-(3-Ethoxy-4-ethoxycarbonylphenyl)acetic acid
2-(3-Ethoxy-4-ethoxycarbonylphenyl)acetic acid
Formel: C₁₃H₁₆O₅
Molecular Weight: 252,27 g/mol
Schmelzpunkt: 80 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08704233
CAS-Nummer: 99469-99-5
EINECS: 427-630-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR