3-Pyridin-4-ylpropionic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(4-Pyridyl)propionic Acid
OR1370-5G
APOSOR1370-5GEA
140
EUR
APOSOR1370-5G
3-Pyridin-4-ylpropionic acid 98%
3-Pyridin-4-ylpropionic acid
Formel:
C₈H₉NO₂ Molecular Weight: 151,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00084846 CAS-Nummer: 6318-43-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...