4-tert-Butyl-3-methoxybenzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(tert-Butyl)-3-methoxybenzoic acid

OR13716-1G OR13716-500MG OR13716-5G
APOSOR13716-1GEA 58 EUR
APOSOR13716-1G APOSOR13716-500MG APOSOR13716-5G
4-tert-Butyl-3-methoxybenzoic acid
4-tert-Butyl-3-methoxybenzoic acid
Formel: C₁₂H₁₆O₃
Schmelzpunkt: 151…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06656116
CAS-Nummer: 79822-46-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR