3-Iod-o-toluylsäure

Lieferant: Apollo Scientific
Achtung

OR13733-100G OR13733-10G OR13733-25G
APOSOR13733-100GEA 144 EUR
APOSOR13733-100G APOSOR13733-10G APOSOR13733-25G
3-Iod-o-toluylsäure
3-Iod-o-toluylsäure
Formel: C₈H₇IO₂
Molecular Weight: 262.05 g/mol
Schmelzpunkt: 124…126 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00270112
CAS-Nummer: 133232-56-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR