2-Amino-4-methoxypyridin
Lieferant: Apollo Scientific
|
Achtung
|
OR13856-5G
OR13856-25G
APOSOR13856-5GEA
39.3
EUR
APOSOR13856-5G
APOSOR13856-25G
2-Amino-4-methoxypyridin
2-Amino-4-methoxypyridine
Formel:
C₆H₈N₂O Molecular Weight: 124,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07437849 CAS-Nummer: 10201-73-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...