2-Bromo-6-hydrazinylpyridine 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromo-6-hydrazinopyridine

OR13857-1G OR13857-25G OR13857-5G
APOSOR13857-1GEA 21.9 EUR
APOSOR13857-1G APOSOR13857-25G APOSOR13857-5G
2-Bromo-6-hydrazinylpyridine 98%
2-Bromo-6-hydrazinylpyridine
Formel: C₅H₆BrN₃
Molecular Weight: 188.028 g/mol
Schmelzpunkt: 117…118 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09743754
CAS-Nummer: 26944-71-8
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR