3-(4-Methylpiperazin-1-yl)benzoic acid

Lieferant: Apollo Scientific
Achtung

OR13862-1G OR13862-25G OR13862-5G
APOSOR13862-1GEA 50.9 EUR
APOSOR13862-1G APOSOR13862-25G APOSOR13862-5G
3-(4-Methylpiperazin-1-yl)benzoic acid
3-(4-Methylpiperazin-1-yl)benzoic acid
Formel: C₁₂H₁₆N₂O₂
Schmelzpunkt: 187…190 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06742238
CAS-Nummer: 215309-01-6
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR