3-(4-Methylpiperazin-1-yl)benzylamine

Lieferant: Apollo Scientific
Gefahr

OR13863-1G OR13863-250MG
APOSOR13863-1GEA 0 EUR
APOSOR13863-1G APOSOR13863-250MG
3-(4-Methylpiperazin-1-yl)benzylamine
3-(4-Methylpiperazin-1-yl)benzylamine
Formel: C₁₂H₁₉N₃
Molecular Weight: 205.31 g/mol
Schmelzpunkt: 78…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368540
CAS-Nummer: 672325-37-0
UN: 2735
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR