5-(4-Methylpiperazino)-2-nitroaniline
Lieferant: Apollo Scientific
Synonyme:
5-(4-Methylpiperazin-1-yl)-2-nitroaniline
OR13864-1G
OR13864-5G
APOSOR13864-1GEA
23.2
EUR
APOSOR13864-1G
APOSOR13864-5G
5-(4-Methylpiperazino)-2-nitroaniline
5-(4-Methylpiperazino)-2-nitroaniline
Formel:
C₁₁H₁₆N₄O₂ Molecular Weight: 236,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00545964 CAS-Nummer: 23491-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...