N-(4-Piperidinyl)acetamid

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(Piperidin-4-yl)acetamide

OR13965-1G OR13965-5G
APOSOR13965-1GEA 69.6 EUR
APOSOR13965-1G APOSOR13965-5G
N-(4-Piperidinyl)acetamid
N-(4-Piperidinyl)acetamid
Formel: C₇H₁₄N₂O
Molecular Weight: 142,20 g/mol
Schmelzpunkt: 141 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05254805
CAS-Nummer: 5810-56-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR