Methyl-4-piperidinylacetathydrochlorid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Piperidyl)acetic Acid Methyl Ester Hydrochloride, Methyl 2-(piperidin-4-yl)acetate hydrochloride
OR13982-25G
APOSOR13982-25GEA
101
EUR
APOSOR13982-25G
Methyl-4-piperidinylacetathydrochlorid
Methyl (4-piperidyl)acetate hydrochloride
Formel:
C₈H₁₅NO₂ Molecular Weight: 193,67 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09261419 CAS-Nummer: 81270-37-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...