4-Cyan-4'-heptylbiphenyl 98%
Lieferant: Apollo Scientific
Synonyme:
4-Cyano-4'-n-heptylbiphenyl, 4'-Heptyl-[1,1'-biphenyl]-4-carbonitrile
OR13990-25G
APOSOR13990-25GEA
95.7
EUR
APOSOR13990-25G
4-Cyan-4'-heptylbiphenyl 98%
4-Cyan-4'-heptylbiphenyl
Formel:
C₂₀H₂₃N Molecular Weight: 277,41 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00074915 CAS-Nummer: 41122-71-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...