4'-Hydroxybiphenyl-4-carbonsäure 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4'-Hydroxy-4-biphenylcarboxylic acid, 4'-Hydroxy-[1,1'-biphenyl]-4-carboxylic acid

OR13999-25G OR13999-5G OR13999-100G
APOSOR13999-25GEA 60.9 EUR
APOSOR13999-25G APOSOR13999-5G APOSOR13999-100G
4'-Hydroxybiphenyl-4-carbonsäure 95%
4'-Hydroxybiphenyl-4-carbonsäure
Formel: C₁₃H₁₀O₃
Molecular Weight: 214.22 g/mol
Schmelzpunkt: 295 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00059078
CAS-Nummer: 58574-03-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR