5-Bromo-1-benzothiophene-3-carboxylic acid 97+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromobenzo[b]thiophene-3-carboxylic acid
OR1401-1G
OR1401-250MG
APOSOR1401-1GEA
0
EUR
APOSOR1401-1G
APOSOR1401-250MG
5-Bromo-1-benzothiophene-3-carboxylic acid 97+%
5-Bromo-1-benzothiophene-3-carboxylic acid
Formel:
C₉H₅BrO₂S Molecular Weight: 257,11 g/mol Schmelzpunkt: 292…293 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797473 CAS-Nummer: 7312-24-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...