4-(5-Methyl-1,2,4-oxadiazol-3-yl)benzoic acid

Lieferant: Apollo Scientific
Achtung

OR1402-1G OR1402-5G
APOSOR1402-1GEA 55.1 EUR
APOSOR1402-1G APOSOR1402-5G
4-(5-Methyl-1,2,4-oxadiazol-3-yl)benzoic acid
4-(5-Methyl-1,2,4-oxadiazol-3-yl)benzoic acid
Formel: C₁₀H₈N₂O₃
Molecular Weight: 204,19 g/mol
Schmelzpunkt: 262…264 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06797475
CAS-Nummer: 95124-68-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR