4-(2,1,3-Benzothiadiazol-5-ylmethoxy)benzenecarboxylic acid

Lieferant: Apollo Scientific

Synonyme: 4-[(2,1,3-Benzothiadiazol-5-yl)methoxy]benzoic acid

OR14029-1G OR14029-500MG
APOSOR14029-1GEA 671 EUR
APOSOR14029-1G APOSOR14029-500MG
4-(2,1,3-Benzothiadiazol-5-ylmethoxy)benzenecarboxylic acid
4-(2,1,3-Benzothiadiazol-5-ylmethoxy)benzenecarboxylic acid
Formel: C₁₄H₁₀N₂O₃S
Molecular Weight: 286.31 g/mol
Schmelzpunkt: 248…249 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06808684
CAS-Nummer: 874834-22-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR