6-Bromo-1-benzothiophene-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Bromobenzo[b]thiophene-2-carboxylic acid
OR14032-1G
OR14032-5G
OR14032-25G
APOSOR14032-1GEA
21.9
EUR
APOSOR14032-1G
APOSOR14032-5G
APOSOR14032-25G
6-Bromo-1-benzothiophene-2-carboxylic acid
6-Bromo-1-benzothiophene-2-carboxylic acid
Formel:
C₉H₅BrO₂S Molecular Weight: 257,11 g/mol Schmelzpunkt: 262…265 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07371540 CAS-Nummer: 19075-58-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...