3-(1,3-Oxazol-5-yl)benzoic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(5-Oxazolyl)benzoic Acid

OR1404-1G OR1404-5G
APOSOR1404-1GEA 194 EUR
APOSOR1404-1G APOSOR1404-5G
3-(1,3-Oxazol-5-yl)benzoic acid 97%
3-(1,3-Oxazol-5-yl)benzoic acid
Formel: C₁₀H₇NO₃
Molecular Weight: 189,17 g/mol
Schmelzpunkt: 258…260 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06797481
CAS-Nummer: 252928-82-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR