tert-Butyl-4-piperazinotetrahydro-1(2H)-pyridinecarboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 4-(piperazin-1-yl)piperidine-1-carboxylate, 4-(Piperazin-1-yl)piperidine, N-BOC protected

OR14041-100MG OR14041-1G OR14041-500MG
APOSOR14041-100MGEA 23.2 EUR
APOSOR14041-100MG APOSOR14041-1G APOSOR14041-500MG
tert-Butyl-4-piperazinotetrahydro-1(2H)-pyridinecarboxylate
tert-Butyl-4-piperazinotetrahydro-1(2H)-pyridinecarboxylate
Formel: C₁₄H₂₇N₃O₂
Molecular Weight: 269,39 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04973999
CAS-Nummer: 177276-41-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR