4-Chlor-2-benzo[b]thiophencarbonsäure

Lieferant: Apollo Scientific
Achtung

OR14042-1G OR14042-5G
APOSOR14042-1GEA 46.4 EUR
APOSOR14042-1G APOSOR14042-5G
4-Chlor-2-benzo[b]thiophencarbonsäure
4-Chlor-2-benzo[b]thiophencarbonsäure
Formel: C₉H₅ClO₂S
Molecular Weight: 212,65 g/mol
Schmelzpunkt: 257…259 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06655155
CAS-Nummer: 23967-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR