2-(4-Ethoxyphenyl)-1,3-thiazolan

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-Ethoxyphenyl)-1,3-thiazolidine

OR14053-10G OR14053-1G OR14053-5G
APOSOR14053-10GEA 894 EUR
APOSOR14053-10G APOSOR14053-1G APOSOR14053-5G
2-(4-Ethoxyphenyl)-1,3-thiazolan
2-(4-Ethoxyphenyl)-1,3-thiazolan
Formel: C₁₁H₁₅NOS
Schmelzpunkt: 96…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027144
CAS-Nummer: 937602-47-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR