Methyl-2-amino-1,3-benzothiazole-6-carboxylate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-amino-1,3-benzothiazole-6-carboxylate

OR14063-1G
APOSOR14063-1GEA 21.9 EUR
APOSOR14063-1G
Methyl-2-amino-1,3-benzothiazole-6-carboxylate 98%
Methyl-2-amino-1,3-benzothiazole-6-carboxylate
Formel: C₉H₈N₂O₂S
Molecular Weight: 208,24 g/mol
Schmelzpunkt: 244…246 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00468672
CAS-Nummer: 66947-92-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR