3-(5-Methyl-1,2,4-oxadiazol-3-yl)benzoic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(5-methyl-1,2,4-oxadiazol-3-yl)benzoic acid

OR1407-1G OR1407-5G
APOSOR1407-1GEA 72.5 EUR
APOSOR1407-1G APOSOR1407-5G
3-(5-Methyl-1,2,4-oxadiazol-3-yl)benzoic acid 97%
3-(5-Methyl-1,2,4-oxadiazol-3-yl)benzoic acid
Formel: C₁₀H₈N₂O₃
Molecular Weight: 204,19 g/mol
Schmelzpunkt: 223…225 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06797485
CAS-Nummer: 264264-32-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR